Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017347739.1 | XP_017203228.1 | 22 - 23 | 3078 | 100 | |
XM_017347740.1 | XP_017203229.1 | 21 - 22 | 3009 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
