Assay Details
Gene Symbol: HTR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708384.3 | XP_002708430.1 | 5 - 6 | 638 | 63 | |
GenBank mRNA | AF305700.1 | - | 1 - 2 | 141 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
