Assay Details
Gene Symbol: AIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717177.3 | XP_002717223.1 | 5 - 6 | 725 | 76 | |
GenBank mRNA | AJ879471.1 | - | 3 - 4 | 476 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
