Assay Details
Gene Symbol: FGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717238.3 | XP_002717284.1 | 2 - 3 | 321 | 150 | |
GenBank mRNA | L12034.1 | - | 2 - 3 | 252 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
