Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195708.1 | NP_001182637.1 | 4 - 5 | 528 | 123 | |
GenBank mRNA | EB372653.1 | - | 4 - 5 | 517 | 123 | |
EB382732.1 | - | 2 - 3 | 150 | 123 | ||
M98395.1 | - | 3 - 4 | 427 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
