Assay Details
Gene Symbol: PROC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195801.1 | NP_001182730.1 | 5 - 6 | 538 | 99 | |
XM_008258416.2 | XP_008256638.1 | 6 - 7 | 659 | 99 | ||
XM_008258417.2 | XP_008256639.1 | 6 - 7 | 664 | 99 | ||
GenBank mRNA | U49933.1 | - | 4 - 5 | 520 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
