Assay Details
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082006.1 | NP_001075475.1 | 1 - 2 | 367 | 138 | |
XM_008261482.2 | XP_008259704.1 | 2 - 3 | 532 | 138 | ||
GenBank mRNA | AB042195.1 | - | 1 - 2 | 367 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
