Assay Details
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082006.1 | NP_001075475.1 | 2 - 3 | 511 | 76 | |
XM_008261482.2 | XP_008259704.1 | 3 - 4 | 676 | 76 | ||
GenBank mRNA | AB042195.1 | - | 2 - 3 | 511 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
