Assay Details
Gene Symbol: ATP1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082091.1 | NP_001075560.1 | 6 - 7 | 680 | 146 | |
GenBank mRNA | AF302929.1 | - | 6 - 7 | 680 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
