Assay Details
Gene Symbol: AK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082114.1 | NP_001075583.1 | 3 - 4 | 445 | 88 | |
GenBank mRNA | AF417508.1 | - | 3 - 4 | 445 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
