Assay Details
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082167.1 | NP_001075636.1 | 125 | 106 | ||
GenBank mRNA | AY553907.1 | - | 125 | 106 |

More Information
Set Membership: |
![]() ![]() |
