Assay Details
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082238.1 | NP_001075707.1 | 6 - 7 | 682 | 67 | |
XM_008257929.2 | XP_008256151.1 | 6 - 7 | 771 | 67 | ||
XM_008257930.2 | XP_008256152.1 | 6 - 7 | 753 | 67 | ||
XM_017342433.1 | XP_017197922.1 | 6 - 7 | 798 | 67 | ||
XM_017342435.1 | XP_017197924.1 | 6 - 7 | 776 | 67 | ||
GenBank mRNA | K02300.1 | - | 6 - 7 | 682 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
