Assay Details
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082261.1 | NP_001075730.1 | 196 | 68 | ||
GenBank mRNA | M11306.1 | - | 196 | 68 |

More Information
Set Membership: |
![]() ![]() |
