Assay Details
Gene Symbol: WARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082288.1 | NP_001075757.1 | 1092 | 102 | ||
XM_008251449.2 | XP_008249671.1 | 1096 | 102 | |||
GenBank mRNA | M33460.1 | - | 1092 | 102 |

More Information
Set Membership: |
![]() ![]() |
