Assay Details
Gene Symbol: RHO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082349.1 | NP_001075818.1 | 3 - 4 | 703 | 122 | |
XM_008260202.2 | XP_008258424.1 | 4 - 5 | 2191 | 122 | ||
GenBank mRNA | U21688.1 | - | 3 - 4 | 703 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
