Assay Details
Gene Symbol: ACE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082395.1 | NP_001075864.1 | 19 - 20 | 2774 | 74 | |
NM_001171069.1 | NP_001164540.1 | 7 - 8 | 1038 | 74 | ||
GenBank mRNA | J05041.1 | - | 7 - 8 | 1038 | 74 | |
X62551.1 | - | 2774 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
