Assay Details
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082655.1 | NP_001076124.1 | 19 - 20 | 2203 | 119 | |
GenBank mRNA | AF388202.1 | - | 19 - 20 | 2203 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
