Assay Details
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082747.1 | NP_001076216.1 | 889 | 104 | ||
GenBank mRNA | S61699.1 | - | 889 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
