Assay Details
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101686.1 | NP_001095156.1 | 1209 | 146 | ||
GenBank mRNA | X06087.1 | - | 1209 | 146 |

More Information
Set Membership: |
![]() ![]() |
