Assay Details
Gene Symbol: LPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101689.1 | NP_001095159.1 | 2 - 3 | 766 | 101 | |
GenBank mRNA | X07995.1 | - | 2 - 3 | 766 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
