Assay Details
Gene Symbol: LPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101689.1 | NP_001095159.1 | 6 - 7 | 1745 | 71 | |
GenBank mRNA | X07995.1 | - | 6 - 7 | 1745 | 71 |
Gene Symbol: LCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712428.3 | XP_002712474.1 | 6 - 7 | 1703 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
