Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105673.1 | NP_001099143.1 | 6 - 7 | 900 | 135 | |
XM_008258588.2 | XP_008256810.1 | 7 - 8 | 1065 | 135 | ||
XM_017341452.1 | XP_017196941.1 | 7 - 8 | 1149 | 135 | ||
GenBank mRNA | EU014879.1 | - | 6 - 7 | 900 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
