Assay Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105676.1 | NP_001099146.1 | 3 - 4 | 420 | 73 | |
GenBank mRNA | EF494239.1 | - | 3 - 4 | 420 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
