Assay Details
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082627.2 | NP_001076096.2 | 5 | 706 | 90 | |
XM_017347219.1 | XP_017202708.1 | 5 | 638 | 90 | ||
GenBank mRNA | L12405.1 | - | 5 | 649 | 90 | |
Z22644.1 | - | 649 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
