Assay Details
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082666.1 | NP_001076135.1 | 1 - 2 | 52 | 78 | |
GenBank mRNA | J05042.1 | - | 1 - 2 | 52 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082666.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
