Assay Details
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082666.1 | NP_001076135.1 | 2 - 3 | 180 | 90 | |
XM_017346640.1 | XP_017202129.1 | 3 - 4 | 285 | 90 | ||
GenBank mRNA | J05042.1 | - | 2 - 3 | 180 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082666.1, XM_017346640.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
