Assay Details
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082770.1 | NP_001076239.1 | 3 - 4 | 345 | 76 | |
GenBank mRNA | D21832.1 | - | 3 - 4 | 336 | 76 | |
M57526.1 | - | 3 - 4 | 345 | 76 | ||
S68977.1 | - | 3 - 4 | 346 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
