Assay Details
Gene Symbol: NPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160286.1 | NP_001153758.1 | 2 - 3 | 275 | 78 | |
GenBank mRNA | AB469827.1 | - | 2 - 3 | 275 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
