Assay Details
Gene Symbol: MIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099961.1 | NP_001093431.1 | 3 - 4 | 641 | 101 | |
GenBank mRNA | EF457887.1 | - | 3 - 4 | 641 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
