Assay Details
Gene Symbol: LOC100328613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171072.1 | NP_001164543.1 | 938 | 74 | ||
XM_008252400.2 | XP_008250622.1 | 951 | 74 | |||
GenBank mRNA | M11727.1 | - | 74 | |||
M19917.1 | - | 74 | ||||
X05685.1 | - | 938 | 74 |

More Information
Set Membership: |
![]() ![]() |
