Assay Details
Gene Symbol: LOC100328613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171072.1 | NP_001164543.1 | 1271 | 71 | ||
XM_008252400.2 | XP_008250622.1 | 1284 | 71 | |||
GenBank mRNA | M11727.1 | - | 963 | 71 | ||
X05685.1 | - | 1271 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
