Assay Details
Gene Symbol: LOC100328937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171121.1 | NP_001164592.1 | 1276 | 76 | ||
XM_008252402.2 | XP_008250624.1 | 1352 | 76 | |||
GenBank mRNA | M11728.1 | - | 973 | 76 | ||
M36538.1 | - | 1276 | 76 | |||
X05686.1 | - | 1295 | 76 | |||
X13853.1 | - | 1277 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
