Assay Details
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204846.1 | NP_001191775.1 | 1 - 2 | 1921 | 116 | |
XM_008252788.2 | XP_008251010.1 | 2 - 3 | 2051 | 116 | ||
XM_008252794.2 | XP_008251016.1 | 2 - 3 | 2085 | 116 | ||
XM_008252798.2 | XP_008251020.1 | 2 - 3 | 2050 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
