Assay Details
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285834.1 | NP_001272763.1 | 1 | 471 | 97 | |
GenBank mRNA | DN882784.1 | - | 1 | 372 | 97 | |
DN883835.1 | - | 1 | 373 | 97 | ||
EL341892.1 | - | 1 | 472 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
