Assay Details
Gene Symbol: QDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319581.1 | NP_001306510.1 | 3 - 4 | 289 | 83 | |
GenBank mRNA | KP247546.1 | - | 3 - 4 | 289 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
