Assay Details
Gene Symbol: QDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319581.1 | NP_001306510.1 | 4 - 5 | 434 | 96 | |
GenBank mRNA | KP247546.1 | - | 4 - 5 | 434 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
