Assay Details
Gene Symbol: QDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319581.1 | NP_001306510.1 | 6 - 7 | 627 | 77 | |
GenBank mRNA | KP247546.1 | - | 6 - 7 | 627 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
