Assay Details
Gene Symbol: LOC100009136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082300.1 | NP_001075769.1 | 246 | 98 | ||
GenBank mRNA | L10842.1 | - | 223 | 98 | ||
M64602.1 | - | 246 | 98 | |||
S55578.1 | - | 223 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
