Assay Details
Gene Symbol: PDCD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723193.3 | XP_002723239.1 | 280 | 58 | ||
XM_008251395.2 | XP_008249617.1 | 281 | 58 | |||
XM_017339218.1 | XP_017194707.1 | 283 | 58 | |||
XM_017339220.1 | XP_017194709.1 | 108 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017339220.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
