Assay Details
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710353.3 | XP_002710399.1 | 4 - 5 | 832 | 68 | |
XM_008255356.2 | XP_008253578.1 | 4 - 5 | 832 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
