Assay Details
Gene Symbol: RSKR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719069.3 | XP_002719115.2 | 6 - 7 | 856 | 79 | |
XM_008271027.2 | XP_008269249.1 | 6 - 7 | 856 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
