Assay Details
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717229.3 | XP_002717275.2 | 3 - 4 | 379 | 97 | |
XM_008267904.2 | XP_008266126.1 | 4 - 5 | 960 | 97 | ||
XM_008267905.2 | XP_008266127.1 | 3 - 4 | 873 | 97 | ||
XM_008267906.2 | XP_008266128.1 | 4 - 5 | 941 | 97 | ||
XM_017347551.1 | XP_017203040.1 | 4 - 5 | 470 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
