Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709690.3 | XP_002709736.1 | 4 - 5 | 1326 | 67 | |
XM_017339318.1 | XP_017194807.1 | 4 - 5 | 1236 | 67 | ||
XM_017339321.1 | XP_017194810.1 | 4 - 5 | 1236 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
