Assay Details
Gene Symbol: PIGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719564.3 | XP_002719610.1 | 3 - 4 | 563 | 74 | |
XM_008271837.2 | XP_008270059.1 | 3 - 4 | 561 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
