Assay Details
Gene Symbol: TPPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717882.2 | XP_002717928.1 | 2 - 3 | 332 | 107 | |
XM_008269322.1 | XP_008267544.1 | 2 - 3 | 322 | 107 | ||
XM_008269323.2 | XP_008267545.1 | 2 - 3 | 317 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
