Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017346018.1 | XP_017201507.1 | 19 - 20 | 2600 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017346018.1 | XP_017201507.1 | 19 - 20 | 2600 | 103 |
Set Membership: |
![]() ![]() ![]() |