Assay Details
Gene Symbol: RIMKLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008265465.1 | XP_008263687.1 | 2 - 3 | 501 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RIMKLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008265465.1 | XP_008263687.1 | 2 - 3 | 501 | 71 |
Set Membership: |
![]() ![]() ![]() |