Assay Details
Gene Symbol: DMAC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718233.3 | XP_002718279.1 | 3 - 4 | 242 | 107 | |
XM_008269635.2 | XP_008267857.1 | 3 - 4 | 341 | 107 | ||
XM_008269636.2 | XP_008267858.1 | 3 - 4 | 224 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
