Assay Details
Gene Symbol: PRSS55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248975.2 | XP_008247197.1 | 4 - 5 | 721 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRSS55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248975.2 | XP_008247197.1 | 4 - 5 | 721 | 92 |
Set Membership: |
![]() ![]() ![]() ![]() |