Assay Details
Gene Symbol: PRSS58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248866.2 | XP_008247088.1 | 107 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008248866.2 transcripts.
Set Membership: |
![]() ![]() |
